Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Glycine Tablet

Catalog No. 09534622 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
100/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-534-622 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-534-622 Supplier Lovibond™ Supplier No. 512170BT
Add to Cart
Add to Cart

Lovibond Glycine Tablet, Range: 0.01 to 3.8mg/L, White, Composition: Non-hazardous components 100%, Format: Tablet

Specifications

Color White
Composition Non-hazardous components 100%
pH Range 5.8
Format Tablet
For Use With (Application) Water analysis, chlorine dioxide aqua comparator™ test kit, ozone aqua comparator™ test kit, chlorine dioxide reagent set, ozone reagent set
For Use With (Equipment) MC500 colorimeter, SP600 spectrophotometer, SC450 colorimeter
Quantity 100/Pk.
Range 0.01 to 3.8mg/L
Storage Requirements 20°C
Test Method DPD
Type Glycine Tablet
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.