Learn More
Lovibond™ Halogen Lamp for SP600 Spectrophotometer
Catalog No. 15398328 Shop All Tintometer Group Products
Change view
Click to view available options
Type:
Replacement Tungsten Halogen Lamp Assembly
Catalog No. Type
15-398-328 Replacement Tungsten Halogen Lamp Assembly
1 options
Catalog No. 15-398-328 Supplier Lovibond™ Supplier No. 711000
Add to Cart
Add to Cart
Replacement tungsten halogen lamp assembly (6V,10w) for SP600 Spectrophotometer.
Replacement tungsten halogen lamp assembly (6V,10w) for SP600 Spectrophotometer.
Specifications
Type Replacement Tungsten Halogen Lamp Assembly
For Use With (Equipment) SP600 spectrophotometer