Learn More
Lovibond™ Legionella Enterprise Test Kit
Catalog No. 15398604 Shop All Tintometer Group Products
Change view
Click to view available options
No. of Tests:
4
Catalog No. No. of Tests
15-398-604 4
1 options
Catalog No. 15-398-604 Supplier Lovibond™ Supplier No. 56B006501
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
The Legionella Enterprise Test Kit provides a combination of Legionella testing methods for the most sensitive and comprehensive evaluation. It allows you to easily collect a filtered water sample and provides the tools to detect Legionella pneumophila serogroup 1 where it grows – in biofilms that coat the surface of pipes, shower heads, air conditioning units, water tanks, taps, spas and other water systems.
This Legionella Test Kit contains filters for a more sensitive test as well as the tools to help pinpoint the source of contamination in a system. Everything you need to collect a sample by swabbing pipes and other surfaces where biofilms are growing is included. This test can provide a rapid indication of the presence of Legionella pneumophila serogroup 1, the most recorded cause of Legionnaires Disease.
With results in 30 to 35 minutes (including sample prep-time), the test allows for immediate understanding and response to the water conditions in a water system. Regular use will increase confidence in your Legionella control procedures and reduce risk of outbreaks and litigation.
  • Ideal for beginning Legionella testing
  • Pinpoint the source of Legionella contamination
  • Confirm performance of Legionella prevention and treatment protocols
  • Convenient onsite testing – no need to transport sample to the lab or send to a specialty lab
  • Respond rapidly to water conditions
Specifications
For Use With (Application) Detect the presence of Legionella pneumophila serogroup 1 bacteria in hot and cold water systems, cooling water, whirlpool spas and in biofilm
Includes 4 test strips with dispensing pipettes, recovery buffer, 2 swabs, 2 single use filters, 2 x 60mL syringes, 2 reaction tubes with recovery buffer and instructions in a carrying case
No. of Tests 4
Test Method Lateral flow immunochromatographic assay
Type Legionella Enterprise Test Kit