Learn More
Lovibond™ Legionella Single Test Kit
Catalog No. 15398603 Shop All Tintometer Group Products
Change view
Click to view available options
No. of Tests:
1
Catalog No. No. of Tests
15-398-603 1
1 options
Catalog No. 15-398-603 Supplier Lovibond™ Supplier No. 56B006601
Add to Cart
Add to Cart
The Legionella Single Test Kit detects the presence of Legionella in a filtered water sample.
This Legionella Test Kit contains everything you need to perform one test with the highest accuracy. With results in 30 to 35 minutes (including sample prep-time), the test allows for immediate understanding and response to the water system conditions. Regular use will increase confidence in your Legionella control procedures and reduce risk of outbreaks and litigation.
  • Accurate, rapid Legionella testing
  • Confirm performance of Legionella prevention and treatment protocols
  • Convenient onsite testing – no need to transport sample to the lab or send to a specialty lab
  • Respond rapidly to the water conditions
Specifications
For Use With (Application) Detecting legionella in a water sample
Includes 1 test strips with dispensing pipette, recovery buffer syringe, sample collection jar, single use filter, 60mL syringe and instructions
No. of Tests 1
Test Method Lateral flow immunochromatographic assay
Type Legionella Single Test Kit