Learn More
Lovibond™ Legionella Swab Test Kit
Catalog No. 15398602 Shop All Tintometer Group Products
Change view
Click to view available options
No. of Tests:
5
Catalog No. No. of Tests
15-398-602 5
1 options
Catalog No. 15-398-602 Supplier Lovibond™ Supplier No. 56B006401
Add to Cart
Add to Cart
The Legionella Swab Test Kit supplies the tools to detect Legionella pneumophila serogroup 1 where it grows – in biofilms that coat the surface of pipes, shower heads, air conditioning units, water tanks, taps, spas and other water systems.
This Legionella Swab Test Kit helps pinpoint the source of contamination in a system. Everything you need to collect a sample by swabbing pipes and other surfaces where biofilms are growing is included. With results in 25 minutes, you'll have confidence that treated water in the water system meets standards and does not pose a public health risk.
  • Pinpoint the source of Legionella contamination
  • Verify performance of Legionella prevention and treatment protocols
  • Convenient onsite testing – no need to transport sample to the lab or send to a specialty lab
Specifications
For Use With (Application) Pinpoint the contamination in a system or to demonstrate cleanliness of an area after a targeted cleaning
Includes 5 test strips, 5 dispensing pipettes, 5 reaction tubes, 5 swabs, 65mL recovery buffer, instruction manual
No. of Tests 5
Test Method Lateral flow immunochromatographic assay
Type Legionella Swab Test Kit