Learn More
Lovibond™ Calibration Set for Infrared Lab Turbidimeter
Catalog No. 09053438 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
1 Set
Catalog No. Quantity
09-053-438 1 Set
1 options
Catalog No. 09-053-438 Supplier Lovibond™ Supplier No. 194150
Add to Cart
Add to Cart
Includes
Standards for 0.1, 20, 200, 800 NTU
For use with TB300-IR Lab Turbidimeter.
For use with TB300-IR Lab Turbidimeter.
Specifications
Type Calibration Kit
For Use With (Equipment) TB 300-IR Lab Turbidimeter
Includes Standards for 0.1, 20, 200, 800 NTU
Quantity 1 Set