Learn More
Description
- Parameter: COD
- Method: Dichromate /H2SO4
Specifications
Specifications
| Color | Yellow to Brown |
| pH Range | 1 |
| Format | Liquid |
| For Use With (Application) | Water quality testing |
| For Use With (Equipment) | MC500 multi-parameter colorimeter |
| No. of Tests | 150 Tests |
| Range | 0 to 1500 mg/L |
| Storage Requirements | 20°C |
| Test Method | Dichromate/H2SO4 |
| Type | COD Tube Test Reagent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.