Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Includes
- Calibrated Sample Container, 100mL
- QAC-Test Tablets (200 tablets)
- Instructions
- Storage Case
Accurate test kits for routine water analysis
- Uses titrimetric method for determination of QAC
- Easy operation and exact reagent dosing
- Foil-wrapped tablet reagents with minimum guaranteed shelf life of five years
- Easy and safe storage
Specifications
Specifications
| For Use With (Application) | Routine water analysis |
| Includes | 100mL calibrated sample container, QAC-test tablets, instructions, storage case |
| No. of Tests | 50 |
| Range | 0 to 80mg/L |
| Type | Quaternary Ammonium Compounds Test Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?