Description
Includes
Four plastic and four glass sample tubes, AC adapter (110V 50/60Hz), four AA batteries, procedure manual and carrying case. Test kits available.
Factory calibrated for over 90 analytes with available test kits (sold separately). Direct-reading digital display. Seven built-in precision wavelength filters cover a range from 420 to 640nm. Colorimeter can also be used as a general-purpose photometer. Wide, uniform 28mm sample tubes (both glass and plastic supplied) with screw caps and 10mL graduations ensure high sensitivity and reproducibility. Reagents are added directly to sample tubes. Colorimeter prompts for insertion of blank tube containing an untreated sample which it uses to autozero and standardize measurement. User is then prompted to insert sample tube and results are displayed in mg/L within seconds. Colorimeter can also display Absorbance and % Transmittance results.
- RS-232-C interface permits printer attachment (available on special order)
- Optional data retention model allows data storage and downloading to a computer
- Performs over 90 prestandardized water tests
- Direct reading results in mg/L, Absorbance and % Transmittance
- With RS-232-C interface
- Low-battery indicator
Warranty and Services
Two year.
Specifications
Specifications
| Type | Model 975MP Portable Analyst |