Learn More
Lovibond™ NC45 Non Contact Benchtop Spectrophotometers: Custom Sample Holder
Catalog No. 09053668 Shop All Tintometer Group Products
Change view
Click to view available options
Type:
Custom Sample Holder
Catalog No. Type
09-053-668 Custom Sample Holder
1 options
Catalog No. 09-053-668 Supplier Lovibond™ Supplier No. 400860
Add to Cart
Add to Cart
Accessory for NC45 benchtop spectrophotometer
It is used for measuring many types of wet and dry samples including powders, pastes, gels, plastics and paints. 'Non-contact' design allows detailed color measurement of irregular shaped items such as granules and pellets.
Specifications
Type Custom Sample Holder
For Use With (Equipment) NC45 benchtop spectrophotometer