Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ PD250 Reagent Dispenser with DPD Free Chlorine Reagent

Catalog No. 15401670 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-401-670 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-401-670 Supplier Lovibond™ Supplier No. 194900
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

DPD Free Chlorine Dispenser and one vial of DPD Free Chlorine Reagent for 250 tests (10mL sample)

Dispenser for Free Chlorine reagent

The PD250 is available for 10mL sample sizes for Free DPD Chlorine Reagent. Simply squeeze the handle and the reagent will be will be released into the sample.

  • Squeezeable handle

Specifications

Volume (Metric) 10 mL
For Use With (Application) Simply squeeze the handle and the chlorine reagent will be will be released into the sample
Includes DPD free chlorine dispenser and DPD free chlorine reagent
Quantity Each
Storage Requirements 10°C to 25°C
Type Free Chlorine Dispenser
Volume (English) 0.002 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.