Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ PD250 Reagent Dispenser with DPD Total Chlorine Reagent

Catalog No. 15398552 Shop All Tintometer Group Products
Change view
Click to view available options
Volume (Metric):
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
15-398-552 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-398-552 Supplier Lovibond™ Supplier No. 194910
Add to Cart
Add to Cart

Includes

DPD Total Chlorine Dispenser and one vial of DPD Total Chlorine Reagent for 250 tests (10mL sample)

Dispenser for Total Chlorine reagent

The PD250 is available for 10mL sample sizes for Total DPD Chlorine Reagent. Simply squeeze the handle and the reagent will be will be released into the sample.

  • Squeezeable handle

Specifications

Volume (Metric) 10 mL
For Use With (Application) Simply squeeze the handle and the chlorine reagent will be will be released into the sample
Includes DPD total chlorine dispenser and DPD total chlorine reagent
Quantity Each
Storage Requirements 10°C to 25°C
Type Total Chlorine Dispenser
Volume (English) 0.002 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.