Learn More
Lovibond™ PD250 Reagent Dispenser Refill: DPD Free Chlorine and Total Chlorine
Catalog No. 09534602 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
250 Tests
Catalog No. Quantity
09-534-602 250 Tests
1 options
Catalog No. 09-534-602 Supplier Lovibond™ Supplier No. 530160
Add to Cart
Add to Cart
For use with SC450 Chlorine and pH colorimeter, MC500 colorimeter and SP600 spectrophotometer
Specifications
For Use With (Application) For chlorine testing
For Use With (Equipment) Used with SC450 chlorine and pH colorimeter, MC500 colorimeter and SP600 spectrophotometer
Includes Includes 1 vial of DPD free chlorine reagent and 1 vial of DPD total chlorine reagent
Quantity 250 Tests
Type Replacement Free and Total Chlorine Reagent