Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Phenol Red Tablet

Catalog No. 09534625 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
100/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-534-625 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-534-625 Supplier Lovibond™ Supplier No. 511790BT
Add to Cart
Add to Cart

Lovibond Phenol Red Tablet, Orange, Composition: Sodium Chloride, Format: Tablet, For Use With: Water analysis, Colorimeter and spectrophotometer, Method: Phenol red, pH Range: 6.5 to 8.4, Shelf Life: 10 Years, Storage Requirements: 20 deg.C

Specifications

Color Orange
Composition Sodium Chloride
pH Range 6.5 to 8.4
Format Tablet
For Use With (Application) Water analysis
For Use With (Equipment) Colorimeter and spectrophotometer
Quantity 100/Pk.
Storage Requirements 20°C
Test Method Phenol red
Type Phenol Red Tablet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.