Learn More
Lovibond™ Phosphate (Ortho) 1 Reagent Ampoule
Catalog No. 09534612 Shop All Tintometer Group Products
Change view
Click to view available options
Range:
5 to 40mg/L
Catalog No. Range
09-534-612 5 to 40mg/L
1 options
Catalog No. 09-534-612 Supplier Lovibond™ Supplier No. 380460
Specifications
For Use With (Application) Water analysis
For Use With (Equipment) MC500 colorimeter
Includes Phosphate (Ortho) 1 ampoules, 25mL sampling tube, ampoule blank
Range 5 to 40mg/L
Test Method Vanadomolybdophosphoric Acid
Type Ortho 1 Reagent