Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Oxyscav Vario Reagent Powder Packs™

Catalog No. 09534465 Shop All Tintometer Group Products
Change view
Click to view available options
Range:
20 to 500μg/L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
09-534-465 20 to 500μg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-534-465 Supplier Lovibond™ Supplier No. 531400
Add to Cart
Add to Cart

Vario reagent for MD600 Colorimeter and SpectroDirect Spectrophotometer

  • Pre-measured
  • Easy Open notch
  • Beveled edge for easy dispense and pour

Specifications

Color Light Yellow
Composition Glycine Iron Sulphate >95% (1:1), Disodium 4,4'-[3-(pyridin-2-yl)-1,2,4-triazine-5,6-diyl]bis(benzenesulphonate) <5%
pH Range 5.9
Format Solid
For Use With (Application) Water analysis, test kit
For Use With (Equipment) MC500 colorimeters and SP600 spectrophotometers
Range 20 to 500μg/L
Type Oxyscav-1 Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.