Learn More
Description
- Simply tear open Powder Pack and pour reagent into sample
- Large design makes them easier to handle
- “Easy Open” notch makes it simple to tear open the package
- Easily create a “V”, to allow user to pour reagent into sample
- Beveled edge prevents reagent from getting stuck in the corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
Specifications
| Certifications/Compliance | US EPA |
| Composition | Dipotassium Hydrogen Orthophosphate 40 to 50%, Potassium Dihydrogenorthophosphate 40 to 50%, Sodium Ascorbate 10 to 20%, 2,2 Bichinolin-4,4'-dicarbon Acid Dipotassium trihydrate 95% |
| Volume (Metric) | 10 mL |
| For Use With (Application) | Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing |
| For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
| Range | 0.05 to 5mg/L |
| Test Method | Bicinchoninate |
| Type | Copper Reagent |
| Volume (English) | 0.33 oz. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.