Learn More
Lovibond™ ECR Reagent Powder Packs™
Catalog No. 09053248 Shop All Tintometer Group Products
Change view
Click to view available options
Range:
0 to 0.22mg/L
Catalog No. Range
09-053-248 0 to 0.22mg/L
1 options
Catalog No. 09-053-248 Supplier Lovibond™ Supplier No. 531620
Add to Cart
Add to Cart
Pre-measured, unit dose reagent for water quaility testing
  • Simply tear open Powder Pack and pour reagent into sample
  • Large design makes them easier to handle
  • “Easy Open” notch makes it simple to tear open the package
  • Easily create a “V”, to allow user to pour reagent into sample
  • Beveled edge prevents reagent from getting stuck in the corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
Certifications/Compliance US EPA
For Use With (Application) Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
For Use With (Equipment) Aluminum colorimeter, colorimeter, spectrophotometer
Range 0 to 0.22mg/L
Test Method Eriochrome Cyanide R
Type ECR Reagent