Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Powder Pack™ Reagents: Iron Ferro; 10mL

Catalog No. 09053228 Shop All Tintometer Group Products
Change view
Click to view available options
Range:
0.02 to 3mg/L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
09-053-228 0.02 to 3mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-053-228 Supplier Lovibond™ Supplier No. 530560
Add to Cart
Add to Cart

Pre-measured, unit dose reagent for water quaility testing

  • Simply tear open Powder Pack and pour reagent into sample
  • Large design makes them easier to handle
  • “Easy Open” notch makes it simple to tear open the package
  • Easily create a “V”, to allow user to pour reagent into sample
  • Beveled edge prevents reagent from getting stuck in the corners

Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing

Compliance

Iron-Ferro reagent is US EPA accepted

Specifications

Certifications/Compliance US EPA
Composition Sodium Dithionite 30 to 40%, Disodium Disulphite 20 to 30%, Sodium Thiosulphate 30 to 40%, 1, 10-phenanthroline Monohydrochloride Monohydrate 1 to 5%, Sodium Citrate tribasic Dihydrate 1 to 10%
Volume (Metric) 10 mL
For Use With (Application) Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Range 0.02 to 3mg/L
Test Method 1,10-Phenanthroline
Type Iron Ferro Reagent
Volume (English) 0.33 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.