Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Ammonia Cyanurate Powder Packs™

Catalog No. 09053243 Shop All Tintometer Group Products
Change view
Click to view available options
Range:
0 to 0.8mg/L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
09-053-243 0 to 0.8mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-053-243 Supplier Lovibond™ Supplier No. 531370
Add to Cart
Add to Cart

Premeasured unit dose reagent for water quality testing

  • Offers same high quality standards as tablet reagents
  • Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
  • Simply tear open Powder Pack and pour reagent into sample
  • Large design makes them easier to handle
  • “Easy Open” notch makes it simple to tear open package
  • Easily create “V”, to allow user to pour reagent into sample
  • Beveled edge prevents reagent from getting stuck in corners

Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing

Specifications

Certifications/Compliance US EPA
Color White
Composition Lithium Hydroxide 1 to 10%, Sodium Dichloroisocyanurate Dihydrate 1 to 5%, Sodium Chloride 80 to 90%
pH Range 12.5
Volume (Metric) 10 mL
Format Powder
For Use With (Application) Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
For Use With (Equipment) Hach™ and other manufacturers test kits, colorimeters and spectrophotometers
Range 0 to 0.8mg/L
Storage Requirements 20°C
Test Method Salicylate
Type Ammonia Cyanurate Reagent
Volume (English) 0.33 oz.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.