Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Powder Packs™ for Free Chlorine

Catalog No. 09053217 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
100 Pack
1000 Pack
Range:
0 to 2 mg/L
0 to 3.5 mg/L
0 to 5 mg/L
Volume (English):
0.16 oz.
0.33 oz.
0.84 oz.
Volume (Metric):
5 mL
10 mL
25 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Range Volume (English) Volume (Metric)
09-053-217 1000 Pack 0 to 3.5 mg/L 0.16 oz. 5 mL
09-053-216 100 Pack 0 to 3.5 mg/L 0.16 oz. 5 mL
09-053-219 1000 Pack 0 to 2 mg/L 0.33 oz. 10 mL
09-053-218 100 Pack 0 to 2 mg/L 0.33 oz. 10 mL
09-053-220 100 Pack 0 to 5 mg/L 0.84 oz. 25 mL
09-053-221 1000 Pack 0 to 5 mg/L 0.84 oz. 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 09-053-217 Supplier Lovibond™ Supplier No. 530093
Only null left
Add to Cart
Add to Cart

Premeasured unit dose DPD reagents for water quality testing

  • For use in SC450™ Colorimeters and Aqua Comparator™ Test Kits
  • Also suitable for use in Hach™ instruments and test kits
  • Easy-to-open, sealed foil packets eliminate risk of cross-contamination
  • Available for 5, 10, and 25mL sample sizes
  • Extended shelf life of up to 5 years

Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing

Compliance

US EPA-accepted DPD method

Specifications

Certifications/Compliance US EPA
Volume (Metric) 5 mL
Format Powder
For Use With (Application) Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
For Use With (Equipment) SC450 colorimeter and Aqua Comparator™ test kits, Hach™ instruments and test kits, spectrophotometer
Quantity 1000 Pack
Range 0 to 3.5 mg/L
Type DPD Free Chlorine Reagent
Volume (English) 0.16 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.