Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Sulfamic Acid Powder Packs™

Catalog No. 09053315 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
100 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-053-315 100 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-053-315 Supplier Lovibond™ Supplier No. 532190
Add to Cart
Add to Cart

Pre-measured, unit dose reagent for water quaility testing

  • Simply tear open Powder Pack and pour reagent into sample
  • Large design makes them easier to handle
  • “Easy Open” notch makes it simple to tear open the package
  • Easily create a “V”, to allow user to pour reagent into sample
  • Beveled edge prevents reagent from getting stuck in the corners

Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing

Specifications

Certifications/Compliance US EPA
Volume (Metric) 10 mL
Format Powder
For Use With (Application) Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
For Use With (Equipment) SC450 colorimeter and Aqua Comparator™ test kits, Hach™ instruments and test kits, spectrophotometer
Quantity 100 Pack
Range 10 to 200mg/L
Type Sulfamic Acid Reagent
Volume (English) 0.33 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.