Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Process Chlorine Analyzer Reagents: Buffer Solution, Free Chlorine

Catalog No. 15399623 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
473 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-399-623 473 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 15-399-623 Supplier Lovibond™ Supplier No. 530223
Add to Cart
Add to Cart
This item is not returnable. View return policy

Formulated to work with Hach™ CL17™ and other manufacturer's online chlorine analyzers

  • Used for 30 day continuous supply

Specifications

Format Liquid
For Use With (Equipment) Hach™ CL17™ process chlorine analyzer
Quantity 473 mL
Type Free Chlorine Buffer Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.