Learn More
Lovibond™ Silica Reagent Set, Powder Packs™, High Range
Catalog No. 09053269 Shop All Tintometer Group Products
Change view
Click to view available options
No. of Tests:
100
Catalog No. No. of Tests
09-053-269 100
1 options
Catalog No. 09-053-269 Supplier Lovibond™ Supplier No. 535700
Add to Cart
Add to Cart
Includes
Silica HR Acid Reagent (10mL sample, 100 pack), Silica HR Citric Acid Reagent (10mL sample, 100 pack), Silica HR Molybdate Reagent (10mL sample, 100 pack)
Premeasured unit dose reagent for water quality testing
  • Offers same high quality standards as tablet reagents
  • Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
  • Simply tear open Powder Pack and pour reagent into sample
  • Large design makes them easier to handle
  • “Easy Open” notch makes it simple to tear open package
  • Easily create “V”, to allow user to pour reagent into sample
  • Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
For Use With (Application) Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
For Use With (Equipment) Hach™ and other manufacturers test kits, colorimeters and spectrophotometers
Includes 10mL silica HR acid reagent (100 pack), 10mL silica HR citric acid reagent (100 pack), 10mL silica HR molybdate reagent (100 pack)
No. of Tests 100
Range 0 to 100mg/L
Test Method Salicylate Silicomolybdate
Type High Range Silica Reagent Set