Learn More
Lovibond™ Ammonia Reagent Set, for Tube Testing, High Range
Catalog No. 09054103 Shop All Tintometer Group Products
Change view
Click to view available options
No. of Tests:
25
Catalog No. No. of Tests
09-054-103 25
1 options
Catalog No. 09-054-103 Supplier Lovibond™ Supplier No. 535650
Add to Cart
Add to Cart
Includes
Ammonia Cyanurate Reagent (5mL sample, 100 pack), Ammonia Salicylate Reagent (5mL , 100 pack), HR AM Dilution Tube (50 ct.)
Premeasured unit dose reagent for water quality testing
  • Offers same high quality standards as tablet reagents
  • Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
  • Simply tear open Powder Pack and pour reagent into sample
  • Large design makes them easier to handle
  • “Easy Open” notch makes it simple to tear open package
  • Easily create “V”, to allow user to pour reagent into sample
  • Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
For Use With (Application) Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
For Use With (Equipment) Hach™ and other manufacturers test kits, colorimeters and spectrophotometers
Includes 5mL ammonia cyanurate reagent (100 pack), 5mL ammonia salicylate reagent (100 pack), 50 ct. HR AM dilution tube
No. of Tests 25
Range 0 to 50mg/L
Test Method Salicylate
Type High Range Ammonia Reagent Set