Learn More
Lovibond™ Nitrate Reagent Set, for Tube Testing, 0 to 30 mg/L Nitrate-Nitrogen
Catalog No. 09053267 Shop All Tintometer Group Products
Change view
Click to view available options
No. of Tests:
25
Catalog No. No. of Tests
09-053-267 25
1 options
Catalog No. 09-053-267 Supplier Lovibond™ Supplier No. 535580
Add to Cart
Add to Cart
Includes
Nitra X Reagent Tube Reagent A (50 ct.), Nitra X Chromotropic Reagent B (50 ct.)
Premeasured unit dose reagent for water quality testing
  • Offers same high quality standards as tablet reagents
  • Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
  • Simply tear open Powder Pack and pour reagent into sample
  • Large design makes them easier to handle
  • “Easy Open” notch makes it simple to tear open package
  • Easily create “V”, to allow user to pour reagent into sample
  • Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
For Use With (Application) Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
For Use With (Equipment) Hach™ and other manufacturers test kits, colorimeters and spectrophotometers
Includes 50 ct. Nitra X reagent tube reagent A, 50 ct. Nitra X chromotropic reagent B
No. of Tests 25
Range 0 to 30mg/L
Test Method Chromotropic Acid
Type Nitrate Tube Test Reagent Set