Learn More
Lovibond™ Reactive Phosphorus Orthophosphate Reagent Set
Catalog No. 09053260 Shop All Tintometer Group Products
Change view
Click to view available options
No. of Tests:
50
Catalog No. No. of Tests
09-053-260 50
1 options
Catalog No. 09-053-260 Supplier Lovibond™ Supplier No. 535200
Add to Cart
Add to Cart
Includes
Phosphate 3 Reagent (10mL sample, 50 pack), Dilution Tube (50 ct.), Deionized water (100mL)
Premeasured unit dose reagent for water quality testing
  • Offers same high quality standards as tablet reagents
  • Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
  • Simply tear open Powder Pack and pour reagent into sample
  • Large design makes them easier to handle
  • “Easy Open” notch makes it simple to tear open package
  • Easily create “V”, to allow user to pour reagent into sample
  • Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
For Use With (Application) Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
For Use With (Equipment) Hach™ and other manufacturers test kits, colorimeters and spectrophotometers
Includes 10mL phosphate 3 reagent (50 pack), 50 ct. dilution tube, 100mL deionized water
No. of Tests 50
Range 0.06 to 5mg/L
Test Method Ascorbic Acid
Type Reactive Phosphorous Reagent Set