Learn More
Lovibond™ Total Nitrogen Reagent Set, for Tube Testing, Low Range
Catalog No. 09053265 Shop All Tintometer Group Products
Change view
Click to view available options
Range:
0 to 25mg/L
Catalog No. Range
09-053-265 0 to 25mg/L
1 options
Catalog No. 09-053-265 Supplier Lovibond™ Supplier No. 535550
Add to Cart
Add to Cart
Includes
Total N Persulfate Reagent, Total Nitrogen Reagent A, Total Nitrogen Reagent B, Total Nitrogen Hydroxide LR Tubes, Total Nitrogen Acid LR/HR Tubes, Deionized Water, 100mL
Premeasured unit dose reagent for water quality testing
  • Offers same high quality standards as tablet reagents
  • Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
  • Simply tear open Powder Pack and pour reagent into sample
  • Large design makes them easier to handle
  • “Easy Open” notch makes it simple to tear open package
  • Easily create “V”, to allow user to pour reagent into sample
  • Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
For Use With (Application) Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
For Use With (Equipment) Hach™ and other manufacturers test kits, colorimeters and spectrophotometers, reactor
Includes Total N persulfate reagent (50 pack), total nitrogen reagent A (50 pack), total nitrogen reagent B (50 pack), 50 ct. total nitrogen hydroxide LR tubes, 50 ct. total nitrogen acid LR/HR tubes, 100mL deionized water
No. of Tests 25
Range 0 to 25mg/L
Test Method Persulfate Digestion
Type Low Range Total Nitrogen Reagent Set