Learn More
Lovibond™ Total Phosphate Reagent Set, for Tube Testing
Catalog No. 09054102 Shop All Tintometer Group Products
Change view
Click to view available options
No. of Tests:
50
Catalog No. No. of Tests
09-054-102 50
1 options
Catalog No. 09-054-102 Supplier Lovibond™ Supplier No. 535210
Add to Cart
Edge
Add to Cart
Includes
Acid Reagent Vial (50ct), Phosphate 3 Reagent (10mL sample, 50 pack), Potassium Persulfate Reagent (10mL sample, 50 pack), Sodium hydroxide (1.54 N, 100mL), Sodium hydroxide (1.00 N, 100mL), Deionized water (100mL)
Premeasured unit dose reagent for water quality testing
  • Offers same high quality standards as tablet reagents
  • Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
  • Simply tear open Powder Pack and pour reagent into sample
  • Large design makes them easier to handle
  • “Easy Open” notch makes it simple to tear open package
  • Easily create “V”, to allow user to pour reagent into sample
  • Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
For Use With (Application) Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
For Use With (Equipment) Hach™ and other manufacturers test kits, colorimeters and spectrophotometers, reactor
Includes 50 ct. acid reagent vial, 10mL phosphate 3 reagent (50 pack), 10mL potassium persulfate reagent (50 pack), 1.54N 100mL sodium hydroxide, 100mL deionized water
No. of Tests 50
Range 0.02 to 1.1mg/L P/0.06 to 3.5mg/L PO4
Test Method Acid digestion/Ascorbic Acid
Type Total Phosphate Reagent Set