Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Replacement Reagents for CHECKIT™ Comparator: Copper No. 2 Tablets

Catalog No. 09053188 Shop All Tintometer Group Products
Change view
Click to view available options
Range:
0.05 to 5, 0 to 1mg/L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
09-053-188 0.05 to 5, 0 to 1mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-053-188 Supplier Lovibond™ Supplier No. 513560BT
Add to Cart
Add to Cart

Specifications

Color Yellow
Composition 20 to 25% Boric acid, 60 to 70% Non-hazardous component, 5 to 10% Calcium Thioglycollate Trihydrate, 2.5% Adipic acid
pH Range 5.7
Format Tablet
For Use With (Application) Water analysis
For Use With (Equipment) Colorimeter and spectrophotometer
Range 0.05 to 5, 0 to 1mg/L
Test Method Biquinoline
Type Copper No. 2 Tablet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.