Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Replacement Reagent for CHECKIT™ Comparator

Catalog No. 09053205 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
100 Pack
100 mL
15 mL
Type:
DEHA Test Solution
EDTA Test Tablets
Format:
Liquid
Tablet
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type Format
09-053-205 100 Pack EDTA Test Tablets Tablet
09-053-200 100 mL DEHA Test Solution Liquid
09-053-199 15 mL DEHA Test Solution Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09-053-205 Supplier Lovibond™ Supplier No. 512390BT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lovibond™ Replacement Reagent for CHECKIT™ Comparator is manufactured under tightly controlled conditions to maintain the highest quality standards.

  • Highest quality production standards
  • Consistent and reliable reagents are provided in liquid and tablet formats
  • Maximum accuracy

Specifications

Color White
Composition Boric Acid 40 to 45%, non-hazardous components 40 to 45%, EDTA Disodium Salt 5 to 15%
pH Range 4.5
Format Tablet
For Use With (Application) Water analysis
For Use With (Equipment) Colorimeter and spectrophotometer
Quantity 100 Pack
Range 0.02 to 0.5mg/L
Type EDTA Test Tablets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.