Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Replacement Reagents for CHECKIT™ Comparator: DEHA Tablets

Catalog No. 09053198 Shop All Tintometer Group Products
Change view
Click to view available options
Range:
20 to 500μg/L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
09-053-198 20 to 500μg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-053-198 Supplier Lovibond™ Supplier No. 513220BT
Add to Cart
Add to Cart

Specifications

Color Yellow
Composition Boric Acid 5 to 10%, non-hazardous components 90 to 100%
pH Range 5.6
Format Tablet
For Use With (Application) Water analysis
For Use With (Equipment) Colorimeter and spectrophotometer
Range 20 to 500μg/L
Storage Requirements 20°C
Test Method PPST
Type DEHA Tablet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.