Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Replacement Reagents for CHECKIT™ Comparator: Phosphate No. 1 LR Tablets

Catalog No. 09053191 Shop All Tintometer Group Products
Change view
Click to view available options
Range:
0.05 to 4mg/L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
09-053-191 0.05 to 4mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-053-191 Supplier Lovibond™ Supplier No. 513040BT
Add to Cart
Add to Cart

Lovibond Replacement Reagents for CHECKIT Comparator: Phosphate No. 1 LR Tablets, Range: 0.05 to 4mg/L, White, Composition: Sodium Bisulfate 50 to 60%, Adipic Acid 10 to 20%, Boric Acid 25 to 35%, Citric Acid 0.1 to 1%, Potassium Antimony Oxide Tartarate Hemi hydrate <0.1%, Format: Tablet

Lovibond Replacement Reagents for CHECKIT Comparator: Phosphate No. 1 LR Tablets, Range: 0.05 to 4mg/L, White, Composition: Sodium Bisulfate 50 to 60%, Adipic Acid 10 to 20%, Boric Acid 25 to 35%, Citric Acid 0.1 to 1%, Potassium Antimony Oxide Tartarate Hemi hydrate <0.1%, Format: Tablet

Specifications

Color White
Composition Sodium Bisulfate 50 to 60%, Adipic Acid 10 to 20%, Boric Acid 25 to 35%, Citric Acid 0.1 to 1%, Potassium Antimony Oxide Tartarate Hemi hydrate <0.1%
pH Range 1.13
Format Tablet
For Use With (Application) Low range phosphate AC test kit, phosphate, ortho LR reagent set, water analysis
For Use With (Equipment) MC500 colorimeters and SP600 spectrophotometers
Range 0.05 to 4mg/L
Storage Requirements 20°C
Test Method Phosphomolybdic Acid/Ascorbic Acid
Type Phosphate No. 1 LR Tablet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.