Learn More
Lovibond™ Replacement Reagents for CHECKIT™ Comparator: Silica No. 2 Tablets
Shop All Tintometer Group ProductsDescription
- These tablet reagents are designed for precise and safe water analysis.
- The tablets are supplied in aluminum blister packs that protect against climatic and environmental conditions to ensure high durability.
- Application: Water analysis
- Parameter and Range: Silica 0.4 to 4.0 mg/L SiO2
- Method No.: M3101
- Hazardous Components: Boric acid
- Key Benefits: Highest accuracy, easy handling, and precise dosing
Specifications
Specifications
| Color | Beige |
| Composition | (+)-Tartaric Acid 40 to 45%, Boric Acid 35 to 40%, non-hazardous components 25 to 35% |
| pH Range | 3.2 |
| Format | Tablet |
| For Use With (Application) | LR silica Aqua comparator™ test kit, silica VLR Aqua comparator™ test kit, silica reagent set, water analysis |
| For Use With (Equipment) | Silica SC450 colorimeter, MC500 colorimeter, SP600 spectrophotometer |
| Range | 0.05 to 4mg/L |
| Test Method | Silicomolybdate |
| Type | Silica No. 2 Tablet |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.