Learn More
Lovibond™ Secondary Reference Standard Kit
Catalog No. 09534620 Shop All Tintometer Group Products
Change view
Click to view available options
For Use With (Equipment):
Chlorine 1 and Chlorine 3 parameter colorimeter
Catalog No. For Use With (Equipment)
09-534-620 Chlorine 1 and Chlorine 3 parameter colorimeter
1 options
Catalog No. 09-534-620 Supplier Lovibond™ Supplier No. 275660
Add to Cart
Add to Cart
Lovibond Secondary Reference Standard Kit, For Use With: Water analysis, Chlorine 1 and Chlorine 3 parameter colorimeter, Includes: Confirms standards of 0, 0.20 and 1mg/L chlorine
Specifications
For Use With (Application) Water analysis
For Use With (Equipment) Chlorine 1 and Chlorine 3 parameter colorimeter
Includes Confirms standards of 0, 0.20 and 1mg/L chlorine
Type Secondary Reference Standard Kit