Learn More
Lovibond™ Glass Filter Set for SpectroDirect & XD7000 Spectrophotometer
Catalog No. 15398324 Shop All Tintometer Group Products
Change view
Click to view available options
Type:
Secondary Standard Set
Catalog No. Type
15-398-324 Secondary Standard Set
1 options
Catalog No. 15-398-324 Supplier Lovibond™ Supplier No. 711160
Add to Cart
Add to Cart
Complete Glass Filter Set for testing the photometric accuracy and the wavelength accuracy of the spectrophotometer
Specifications
Type Secondary Standard Set
For Use With (Equipment) SP600 spectrophotometer
Includes 4 Glass filters for wavelength confirmation