Learn More
Lovibond™ Tablet Reagent: Hardness BW
Catalog No. 09054306 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
250
Catalog No. Quantity
09-054-306 250
1 options
Catalog No. 09-054-306 Supplier Lovibond™ Supplier No. 515171BT
Add to Cart
Add to Cart
Guarantees exact reagent dosing thereby allowing for more precise titrations
  • Convenient blister packaging allows user to push tablet through foil directly into sample
  • Each tablet is hermetically sealed in an individual aluminum foil pocket with shelf life of five years
  • Use with LR Total Hardness MINIKIT
Specifications
For Use With (Application) Routine water analysis
For Use With (Equipment) LR total hardness minikit
Quantity 250
Type Hardness BW Tablet