Learn More
Lovibond™ Molybdate Test Kit, 0 to 100 mg/L
Catalog No. 15398524 Shop All Tintometer Group Products
Change view
Click to view available options
No. of Tests:
100
Catalog No. No. of Tests
15-398-524 100
1 options
Catalog No. 15-398-524 Supplier Lovibond™ Supplier No. 147290
Add to Cart
Add to Cart
Premeasured unit dose reagent for water quality testing
  • Offers same high quality standards as tablet reagents
  • Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
  • Simply tear open Powder Pack and pour reagent into sample
  • Large design makes them easier to handle
  • “Easy Open” notch makes it simple to tear open package
  • Easily create “V”, to allow user to pour reagent into sample
  • Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
For Use With (Application) Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
For Use With (Equipment) Hach™ and other manufacturers test kits, colorimeters and spectrophotometers
Includes Aqua comparator™ housing, aqua comparator™ disc, molybdate No. 1 HR tablets (100 pack), molybdate No. 2 HR tablets (100 pack), 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case
No. of Tests 100
Range 0 to 100mg/L
Type Molybdate Aqua Comparator™ Test Kit