Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Universal pH Tablet

Catalog No. 09534621 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
100/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-534-621 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-534-621 Supplier Lovibond™ Supplier No. 515440
Add to Cart
Add to Cart

Specifications

Composition Cellulose ≤2.5%, Potassium Chloride 90 to 100%, Phenolphthalein ≤0.1%
pH Range 4 to 10
For Use With (Application) Water analysis
For Use With (Equipment) Colorimeter and spectrophotometer
Quantity 100/Pk.
Range 4 to 10 pH
Type Universal pH Tablet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.