Learn More
FENWAL INC Lovo Pouch 6/CS
Catalog No. 501961848
Encompass
This item has a minimum qty of 3 per supplier requirements.
For use with Lovo version 3.0 software and product code X6R4909 8 mL pouch swabable, needleless female luer site and male luer with cap For reagent addition and/or sampling during cell processing Sterile fluid path Individually packaged in plastic overwraps
Catalog No. 50-196-1848 Supplier FENWAL INC Supplier No. X6R4907
This item has a minimum qty of 3 per supplier requirements.
For use with Lovo version 3.0 software and product code X6R4909 8 mL pouch swabable, needleless female luer site and male luer with cap For reagent addition and/or sampling during cell processing Sterile fluid path Individually packaged in plastic overwraps