Learn More
Braintree Scientific LoVol Port, Mini Port only- attaches to a 3F catheter
SDP
Catalog No. 501955096
Encompass
LoVol™ PortUltra-low dead volume port for CSF infusion and withdrawal Ultra-low dead volume of Lovol Port is the port of choice for any application requiring minimal dead volume, i.e. an unusually expensive test article of CSF collection. Available in 2 sizes, MIN-LoVol and MID-LoVol, with reservoir volume only 10% of the Solomon MIN and MID standard SoloPorts. LoVol ports can be accessed with standard 22ga and 24ga huber needles and infusion sets. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5096 Supplier Braintree Scientific Supplier No. MINLOAC30
LoVol™ PortUltra-low dead volume port for CSF infusion and withdrawal Ultra-low dead volume of Lovol Port is the port of choice for any application requiring minimal dead volume, i.e. an unusually expensive test article of CSF collection. Available in 2 sizes, MIN-LoVol and MID-LoVol, with reservoir volume only 10% of the Solomon MIN and MID standard SoloPorts. LoVol ports can be accessed with standard 22ga and 24ga huber needles and infusion sets. **Sold for research purposes only and not for human use**