Learn More
STEMCELL Technologies 35 mm Culture Dishes, Size: 10 Units
SDP
Catalog No. NC9663173
Encompass
35 mm culture dishes, pre-tested for optimal colony growth without supporting anchorage-dependent cells in methylcellulose-based assays. They are manufactured from optical quality, thermally resistant, non-pyrogenic, non-toxic polystyrene.Recommended for:• Hematopoietic progenitor cultures in methylcellulose-based media (MethoCult™)• Embryoid body formation from mouse embryonic and induced pluripotent stem cells• Performing the neural colony-forming cell assay (NCFC)
This item is not returnable. View return policy
Catalog No. NC9663173 Supplier STEMCELL Technologies Supplier No. 27100
35 mm culture dishes, pre-tested for optimal colony growth without supporting anchorage-dependent cells in methylcellulose-based assays. They are manufactured from optical quality, thermally resistant, non-pyrogenic, non-toxic polystyrene.Recommended for:• Hematopoietic progenitor cultures in methylcellulose-based media (MethoCult™)• Embryoid body formation from mouse embryonic and induced pluripotent stem cells• Performing the neural colony-forming cell assay (NCFC)