Learn More
Grainger Rubbermaid Commercial Products Milano Sensor Faucet, Polished Chrome, 8 in. Centers, 1 ea.
Catalog No. 502782247
Encompass_Preferred
The Milano Auto Faucet is a deck-mounted, touch-free automatic faucet designed for commercial washrooms. Utilizing patented Surround Sensor™ Technology, it provides omnidirectional sensing for reliable activation. The system helps maintain hygiene by reducing cross-contamination and includes a built-in mixing valve for adjustable water temperature control.
  • Touch-free activation helps reduce the spread of germs
  • Omnidirectional sensing zone ensures consistent performance
  • Dry Cam-Gear Technology eliminates solenoid valves and reduces maintenance
  • Auto-open line purge feature prevents stagnant water and bacterial growth
  • Battery-powered operation lasts more than 3 years or 400,000 cycles
  • Includes hot and cold supply lines and mixing valve
  • Low lead and ADA compliant design
Catalog No. 50-278-2247 Supplier Grainger Supplier No. 19T381
The Milano Auto Faucet is a deck-mounted, touch-free automatic faucet designed for commercial washrooms. Utilizing patented Surround Sensor™ Technology, it provides omnidirectional sensing for reliable activation. The system helps maintain hygiene by reducing cross-contamination and includes a built-in mixing valve for adjustable water temperature control.
  • Touch-free activation helps reduce the spread of germs
  • Omnidirectional sensing zone ensures consistent performance
  • Dry Cam-Gear Technology eliminates solenoid valves and reduces maintenance
  • Auto-open line purge feature prevents stagnant water and bacterial growth
  • Battery-powered operation lasts more than 3 years or 400,000 cycles
  • Includes hot and cold supply lines and mixing valve
  • Low lead and ADA compliant design