Learn More
Grainger Rubbermaid Commercial Products Milano Auto Faucet, Touch-Free, Metal, 0.5 GPM, 1 ea
Catalog No. 502782003
Encompass_Preferred
The Milano Auto Faucet is an automatic, touch-free bathroom sink faucet engineered for hygiene and efficiency. It utilizes Surround Sensor™ Technology to dispense water only when necessary, facilitating significant water savings. Constructed from durable metal with a polished chrome finish, this faucet is designed for high-traffic environments.
  • Touch-free activation reduces cross-contamination.
  • Omnidirectional sensing zone.
  • Patented Dry Cam-Gear Technology controls water flow and eliminates solenoid valves to prevent repairs.
  • Battery life lasts more than 3 years or 400,000 handwashing cycles (4 D batteries included).
  • Auto-open line purge feature eliminates stagnant water.
  • NSF/ANSI/CAN 61 q ≤ 1 compliant, IAPMO certified, ADA compliant, and low lead compliant.
  • Includes spout, valve control module, mixing valve, and hot/cold supply hoses.
Catalog No. 50-278-2003 Supplier Grainger Supplier No. 19T380
The Milano Auto Faucet is an automatic, touch-free bathroom sink faucet engineered for hygiene and efficiency. It utilizes Surround Sensor™ Technology to dispense water only when necessary, facilitating significant water savings. Constructed from durable metal with a polished chrome finish, this faucet is designed for high-traffic environments.
  • Touch-free activation reduces cross-contamination.
  • Omnidirectional sensing zone.
  • Patented Dry Cam-Gear Technology controls water flow and eliminates solenoid valves to prevent repairs.
  • Battery life lasts more than 3 years or 400,000 handwashing cycles (4 D batteries included).
  • Auto-open line purge feature eliminates stagnant water.
  • NSF/ANSI/CAN 61 q ≤ 1 compliant, IAPMO certified, ADA compliant, and low lead compliant.
  • Includes spout, valve control module, mixing valve, and hot/cold supply hoses.