Learn More
Cole-Parmer Essentials Plus Griffin Low-Form Beaker, Glass, 3000 mL
Catalog No. NC1887609 Shop All Cole-Parmer Products
Encompass_Preferred
These low-form beakers are constructed of Class A borosilicate glass to meet ASTM E960 specifications.
Catalog No. NC1887609 Supplier Cole-Parmer Supplier No. 3450336
These low-form beakers are constructed of Class A borosilicate glass to meet ASTM E960 specifications.