Learn More
Greiner Bio-One Low birefringent (LBR) ComboPlate™ Crystallization Microplates
Catalog No. 07000679 Shop All Greiner Bio-One Products
Change view
Click to view available options
Well Shape:
Flat Bottom (F-well)
Catalog No. Well Shape
07-000-679 Flat Bottom (F-well)
1 options
Catalog No. 07-000-679 Supplier Greiner Bio-One Supplier No. 662850
For superior optical properties with polarized light. Greiner Bio-One Low birefringent (LBR) ComboPlate™ Crystallization Microplates are used for protein crystallization.
  • Virgin, high-clarity polystyrene combined with distortion-free bottom provides optical properties for microscopic inspection of crystal growth
  • Low birefringent (LBR) material for superior optical properties with polarized light
TRUSTED_SUSTAINABILITY
Specifications
Lid With Lid
Array 4 x 6
Color Clear
Material Polystyrene
No. of Wells 24
Sterility Non-sterile
Well Shape Flat Bottom (F-well)
No. per Case 24
Quantity 24/Cs.
Height (Metric) 18.8 mm
Outside Diameter (Metric) Well: 16.3 mm