Learn More
Analytical Sales & Services Low-Carry Modified Hamilton Model 1710N 100µL CTC Syringe, Cemented Needle
Catalog No. 5000804428
Encompass
Low-Carry Modified Hamilton Model 1710N 100µL CTC Syringe w Cemented Needle. 22s Gauge, 51mm Length, Point Style #3\n\nQty/Pkg: EACH
Catalog No. 50-008-04428 Supplier Analytical Sales & Services Supplier No. 4203077014S
Low-Carry Modified Hamilton Model 1710N 100µL CTC Syringe w Cemented Needle. 22s Gauge, 51mm Length, Point Style #3\n\nQty/Pkg: EACH