Learn More
Active Motif Low Cell ChIP Kit 16 rxns
SDP
Catalog No. NC2586639
Encompass
Active Motif's Low Cell ChIP-Seq Kit provides a complete ChIP-Seq workflow for chromatin preparation, immunoprecipitation and NGS library preparation from limited amounts of cells or small tissue biopsies. The assay is robust enough to work with both high abundance histone modifications and low abundance transcription factor proteins. The Next Gen DNA Library and Next Gen DNA Indexing reagents are included so you can generate high complexity sequencing libraries that are compatible with Illumina platforms.
Catalog No. NC2586639 Supplier Active Motif Supplier No. 53086
Active Motif's Low Cell ChIP-Seq Kit provides a complete ChIP-Seq workflow for chromatin preparation, immunoprecipitation and NGS library preparation from limited amounts of cells or small tissue biopsies. The assay is robust enough to work with both high abundance histone modifications and low abundance transcription factor proteins. The Next Gen DNA Library and Next Gen DNA Indexing reagents are included so you can generate high complexity sequencing libraries that are compatible with Illumina platforms.