Learn More
Fine Science Tools High Temperature Cautery Kit, 16 cm, 70 g
SDP
Catalog No. NC9721806
Encompass
Product: Low Cost Cauterizer Kit/ Handle Weight : 60g/ Handle Length : 17.5cm/ Autoclave Safe: No/ Unit : Each
Catalog No. NC9721806 Supplier Fine Science Tools Supplier No. 1801000
Product: Low Cost Cauterizer Kit/ Handle Weight : 60g/ Handle Length : 17.5cm/ Autoclave Safe: No/ Unit : Each