Learn More
Lascar Electronics Inc Low Cost Temperature Data Logger with USB
SDP
Catalog No. NC1856604
Encompass_Preferred
This standalone data logger measures temperature every 30 minutes over a -10 to +50°C (+14 to +122°F) range. Easily set up the logger and view downloaded data by plugging the unit into a PC’s USB port and using the free EasyLog software provided. Data can then be graphed, printed and exported to other applications for detailed analysis. Supplied with a CR1620 battery.
Catalog No. NC1856604 Supplier Lascar Electronics Inc Supplier No. ELUSBLITE
This standalone data logger measures temperature every 30 minutes over a -10 to +50°C (+14 to +122°F) range. Easily set up the logger and view downloaded data by plugging the unit into a PC’s USB port and using the free EasyLog software provided. Data can then be graphed, printed and exported to other applications for detailed analysis. Supplied with a CR1620 battery.